Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
High vacuum bellow valves, VE series
Supplier: VACUUBRAND
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20664004 20664005 20664006 20674020 20674021 20674022
VACU20664004EA 0 INR InStock
VACU20664004 VACU20664005 VACU20664006 VACU20674020 VACU20674021 VACU20674022
High vacuum bellow valves, VE series
Valves Vacuum Valves
High vacuum, helium leak-tested right angle valves VE have stainless steel bellows which seals the drive mechanism without any gap on the vacuum side.
  • High conductance due to large stroke length
  • Excellent gas tightness, minimum degassing
  • Stainless steel version VE WIG bakeable to 150 °C
  • Simple dismantling and easy changing of seal
  • Dimensions of the VE valve are the same as elbow with corresponding nominal width
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms