Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

VWR® Square Polycarbonate Media Bottles

Supplier: VWR Collection

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWRI210-000090EA 0 INR InStock
VWRI210-000090 VWRI210-000094 VWRI210-000102 VWRI210-000095 VWRI210-000101 VWRI210-000103 VWRI210-000100 VWRI210-000093 VWRI210-000099 VWRI210-000096 VWRI210-000098 VWRI210-000087 VWRI210-000097 VWRI210-000104 VWRI210-000089 VWRI210-000086 VWRI210-000091 VWRI210-000088 VWRI210-000092 VWRI210-000085
VWR® Square Polycarbonate Media Bottles
Bottles Plastic Bottles
VWR® square polycarbonate bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −130 to +121 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.

  • Made from durable 100% virgin polycarbonate material
  • Suitable for frozen storage down to −130 °C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and closures (HDPE/PP) provided for applicable products
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-
More Product Information
Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms