Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Vortex Mixer
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
10153-840 10153-810 10153-836
VWRU10153-840EA 0 INR InStock
VWRU10153-840 VWRU10153-810 VWRU10153-836
VWR®, Vortex Mixer
Shakers Vortex Shakers
These vortex mixers are ideal for applications that demand repeatable results.
  • Cup head and three inch platform provided
  • LED display provided
  • Mixer speed: 3200 rpm
The control allows low rpm starts for gentle or high-speed mixing for vigorous vortexing of samples. The mixers have two modes of operation: Continuous mode when using optional accessory attachments or touch mode that activates mixing when the cup head is depressed. The microprocessor control maintains a set speed for strong, consistent mixing action. If users program the timer to a definedMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms