Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Talon®, Retort rings, open
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
28641-012 28641-010
VWRU28641-012EA 0 INR InStock
VWRU28641-012 VWRU28641-010
VWR® Talon®, Retort rings, open
Retort Rings
These lightweight rings mount funnels, boiling flasks, and other irregular-shaped objects to lab frames.
  • Open ring section allows items to pass in and out of support area easily, reducing risk of breakage
  • Mount to rods and frames with connectors such as VWR® Talon® regular or heavy-duty holders
Extension arm length: 57 mm
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms