VWR®, Manual Cell Counter
Supplier: VWR BRAND PRIVATE LABEL USA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
77776-674
VWRU77776-674EA 0 INR
VWRU77776-674
VWR®, Manual Cell Counter
Cell Counters and Flow Cytometers Manual Cell Counters
Light weight digital cell counter water-resistant, O-ring-sealed and shock-resistant ABS plastic case which is ideal for lab and plant use.
  • Easy-to-read LCD
  • Tactile click buttons provide positive action
  • Fail-safe design eliminates accidental resetting
  • Solid construction eliminate gear and miscount problems associated with mechanical counters
Weight: 28,3 g