VWR®, Autoclave deodorant
Supplier: VWR BRAND PRIVATE LABEL USA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
80080-888 80080-896
VWRU80080-888EA 0 INR
VWRU80080-888 VWRU80080-896
VWR®, Autoclave deodorant
Deodorants and Air Fresheners
These steriliser deodorisers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them. Simply place one capsule in your autoclave or incubator with each load. The heat and moisture will dissolve the capsule and release a clean fresh scent.
  • Easy to use
  • Different fragrances available