Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Beakers, Low Form, Borosilicate Glass, Double Scale
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
89000-198
VWRU89000-198EA 0 INR InStock
VWRU89000-198
VWR®, Beakers, Low Form, Borosilicate Glass, Double Scale
Beakers
Borosilicate glass 3.3.
  • With uniform wall thickness throughout
  • All sizes have a durable matte finish marking area and a white graduated scale for measuring and/or mixing where a high level of accuracy is not required
  • Very good chemical resistance, very good temperature resistance
These Griffin beakers feature thick, slightly flared, beaded tops with a spout designed for excellent pouring characteristics.
Certifications: Designed from ASTM Specification E960, Type I requirements.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms