Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Bench Protectors with Leakproof Moisture Barrier
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
89126-794 89126-790 89126-792
VWRU89126-794EA 0 INR InStock
VWRU89126-794 VWRU89126-790 VWRU89126-792
VWR®, Bench Protectors with Leakproof Moisture Barrier
Surface Protectors Bench Protectors
These super absorbent bench protectors feature a two-layer, cushioned construction for use in a laboratory or veterinary setting. The air-lay cellulose layer provides cushion for laboratory equipment and glassware to prevent breakage, while the leakproof plastic backsheet protects tables and benches from spills and moisture.
  • Two layers of protection and leakproof backsheet
  • Available as rolls or mats
  • Can be cut to desired size
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms