Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® sympHony™, Multi-parameter meters, handheld
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
89231-668
VWRU89231-668EA 0 INR InStock
VWRU89231-668
VWR® sympHony™, Multi-parameter meters, handheld
Electrochemistry Meters pH and Multi-Parameter Meters
Specially designed for use in environmental field applications and in the lab, these rugged and durable handheld meters are used with probes to measure various parameters in solutions.
  • Simple to operate, intuitive user interface with step-by-step on screen prompts
  • Consistently accurate and reliable measurements
  • Configurable to each user's unique needs
  • Waterproof casing (IP67 certified)
Each meter features one single connector that can be used when testing for multiple parameters, and also allows for automatic or manual calibration and automatic bufferMore Product Information
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms