Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Electrode filling solution
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
89236-504
VWRU89236-504EA 0 INR InStock
VWRU89236-504
VWR®, Electrode filling solution
Electrolytes Electrode Filling Solutions
This 3M KCl filling solution for appropriate sympHony® Ag/AgCl reference elements.
  • Compatible with waterproof electrodes
  • Easy to dispense
  • Reliable internal reference
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms