Mixing Systems
Combining quality and expertise for all mixing needs
Closed and open systems for powder and liquid mixing applications.
As an industry leader in mixing solutions, Avantor offers unmatched expertise and capability to provide mixing system solutions that work. Whether your operation incorporates a simple liquid-liquid application or more complex liquid-solid applications, Avantor’s suite of open top or closed single-use mixing systems can meet your most challenging mixing demands. As the only open-architecture fluid handling provider, Avantor integrates various single-use componentry from our extensive brand-agnostic portfolio that has been rigorously validated according to Avantor’s best-in-class quality systems.
It’s not just our ability to custom design systems reliably and quickly that sets us apart, but also our decades of experience with fluid transfer systems. Avantor single-use mixing solutions afford our customers peace of mind as they benefit from streamlined workflows, reduced contamination risk, and improved resource efficiency. Our systems are flexible, scalable and configurable to precisely match your production requirements and they can be integrated directly into your production control systems. Our regional technical team members offer expert design, acceptance testing and install support either on-site or remote as needed.
Durable and easy to use with innovative UI across platforms
1
Intuitive software controlled by a 7” color touchscreen display with programmable capability allowing the user to save mixing profiles
2
Integrated loadcell
3
Temperature controlled jacket
Masterflex L/S MasterSense Peristaltic Pump Drive Features
4
Sturdy stainless steel frame with locking casters for ease of positioning
5
Door windows for visual indication of vessel contents
Magnetic Mixing System
The Magnetic Mixing bag offers a sterile, single-use mixing container designed to maximize mixing performance while minimizing disruptive shear. The magnetically driven single use 5-bladed impeller is floating freely by magnetic forces minimizing risk of particle generation and ensuring a safe and homogenous mixing of product. Avantor's magnetic mixing bags come in standard designs ranging, from 100l to 1500l or Avantor's experienced team of engineers can help you design specialized solutions tailored to your unique bioprocessing mixing needs.
Quattro Mixer
Ensure the quality of your product by reducing risk of contamination with an integrated single-use closed mixing system. Avantor’s Quattro Mixer system can process liquids or powders simply and efficiently while designed for current good manufacturing practice (cGMP).
Top Mixer
These modular open top mixing systems can process liquids or powders simply and efficiently while maintaining the highest level of dependability. The Avantor Top Mixer systems can be customized to meet your unique manufacturing specifications, from platform to accessories, compatible with Avantor’s full suite of single-use solutions, including tank liners and other accessories, can meet your production demand.
Click here for more information on the different impeller options for our open top mixing systems.
Unparalleled quality you can rely on
ISO certified cleanrooms
Avantor Fluid Handling assemblies are manufactured in an ISO Class 7 (Class 10000) cleanroom.
Certified, compliant & registered
The assembly facility is IS0 -13485 certified and cGMP compliant.
Product testing and validation
Our operations Incorporate multiple product Inspection and verification steps. Irradiation. endotoxin. particu­late and other testing standards available upon request.
Lot traceability
We offer full lot traceability. Identifying the production site, manufacturing date and origin of components.
3D Bioprocessing bag support containers
3D Bioprocessing Bag Support Containers
Collapsible, stackable and reusable containers to store and transport critical bioprocessing media safely and securely.
Single use container
Interchangeable and reusable open top containers to scale up or scale down product production.