Dispensette® S and Dispensette® S Organic
171 Hero Spot
Safe dispensing of aggressive chemicals
Dispensette® S
Dispensette® S supports a very wide range of applications for the dispensing of aggressive reagents such as concentrated acids and bases
Additional Information
Dispensette® S Organic
Dispensette® S Organic is ideal for dispensing organic solvents.
Additional Information
BRAND has been a pioneer in bottle-top dispensers since 1968 and the Dispensette® is recognised as the leading product in the market. The latest generation Dispensette® S offers new features for safe operation and proven durability for tough operating and media conditions.
Safety features
- Discharge valve with safety ball closes when discharge tube is not mounted
- Filling valve: Olive-shaped for firmer filling tube attachment
- Hinged screw cap doesn't get in the way when dispensing
- The New discharge tube is easy to fasten and is available with or without a recirculation valve
- Optional flexible discharge tube, long and expandable, facilitates fast and precise serial dispensing even into narrow test tubes.
Safety Information - Handling of hazardous substances
Safety information - handling of hazardous substances and many other useful pieces of information are available in the Volumetric Measurement technical handbook.