Cooling Incubators
Featured Products
Cooled incubators with Peltier technology, INCU-Line® ILPR series
Cooled incubators with Peltier technology feature excellent temperature stability and uniformity. They are energy efficient and environmentally friendly, with a low noise level, and they work almost vibration-free as no compressor is installed… save money and save our planet!
    Learn more