Freeze Drying
Freeze Drying
Similar to vacuum concentration, the process of lyophilisation goes one step further by lowering sample temperature to the point where the solution freezes and solvents are removed by sublimation. Designed to handle all the lyophilisation needs of research laboratories.
Best Solutions
Freeze drying systems, FreeZone®
Collectors that reach −50 °C are available with capacities from 1 to 18 litres. Cascade systems in 2, 5, 4, 5, 6 and 12 litre collector capacities that reach −84 °C.
CHECK IT OUT