Laminar Air Flow Systems
Laminar Air Flow Systems
Laminar flow systems are carefully designed to protect a very sensitive products from external contamination. This very clean working atmosphere is obtained by blowing air through HEPA filters. This air stream could be air vertical or horizontal.
Product selection