VWR Ergonomic Chairs for Laboratory and Industry
Human ‘evolution’ is getting worse - make sure that you try to make the best of it!
Laboratory and office work is a striking example of physically awkward sitting positions that can cause blood circulation issues and excessive strain on the body, resulting in fatigue, circulatory disorders, and problems with posture or the musculoskeletal system. We can`t change the evolution of human work but we think that we should do our best to sit you on a chair/stool which helps you stay healthy.
Sitting for prolonged periods in front of your microscope, pipetting at your bench, or just typing stuff on your PC? Then choose from the models below - they will make you happy AND stay healthy.
Click our seating options and browse about YOUR next chair
Reduce spinal stress with our professional level chairs
Having one of these chairs is like visiting a health farm every day! It took 2 years' of study with ergonomic experts, osteopaths and designers to create this range.
The chair is chemical- (weak acids) and rip-resistant, but the biggest benefits are to your wellbeing. This seating is auto-adaptive, so no adjustment is needed plus body weight is managed evenly, so there is a reduction of pressure under the leg which helps to avoid that heavy leg feeling and ensure good circulation. Rebalance your body with the professional! 
    Click for your health
    Find Helpful Resources
    Trailblazing NEON industry chairs
    These chairs combine ergonomics and functional characteristics with a variety of upholstery options, including the super-comfortable SUPERTEC fabric.
    NEON chairs are designed for busy users and have sustainability and economy built in – if the seat upholstery wears out, it can be replaced without having to replace the whole chair!
      Learn more
      Find Helpful Resources
      Certified seating for cleanrooms
      Cleanrooms and sterile environments in microelectronics, pharmaceutical or medical engineering industries present unique seating challenges. We have worked together with industry and research specialists to develop this innovative range.
      Our cleanroom chairs combine comfort whilst meeting the requirements of air cleanliness classification 3 in accordance with DIN EN ISO 14644-1, cleanroom classification 1 as defined by US Federal Standard 209E, and EU GMP guideline specifications.
        Be sure!
        Find Helpful Resources
        Discharge resistance from ESD chairs
        ESD Basic is a tried and tested all-rounder for use in sensitive areas. These low cost chairs provide excellent value for money with an ergonomic design for user comfort and a reliable protection system that ensure discharge resistance in accordance with EN 61340-5-1.
        Experience this high performance workplace chair for the electronics industry and stay grounded!
          Stay safe!
          Find Helpful Resources
          Request a Quote
          Watch More Videos!
          VWR Blue Line The Perfect Setting
          VWR Blue Line Where Is My Chair
          Bimos - Industrial ergonomics of Neon
          Bimos - Changeable upholstery in the Neon 1 + 1 system
          No need to sit for hours? Check out these quick and healthy up and down solutions.
          Basic stools, Mr. Lab range
          Laboratory stool, black artificial leather seat, adjustable in height by pneumatic spring.
          Learn more
          Basic stools, Mr. Lab range - extra wide
          Laboratory stool with extra wide seat in black skai PU foam, PA base (plastic and black steel).
          Learn more
          Basic laboratory stools, Blue Line range
          Laboratory stool with large seating surface, the seat upholstery is made from black PU-foam.
          Learn more
          Scientifically proven ergonomic laboratory stools
          Laboratory stool, PU foam, black.
          Learn more
          Stools, for cleanrooms
          Meet requirements of cleanliness classification 3 in accordance with DIN EN ISO 146444-1, cleanroom classification 1 as defined by US Federal Standard 209E.
          Learn more
          Request a Quote