Masterflex® B/T® Series Peristaltic Pumps
Bulk transfer at its best
Highest flow rate
Flow range: 0.3 to 42 LPM
Makes the job efficient
Masterflex® B/T Series peristaltic pumps have the highest flow rates of any Masterflex® pump. Their benefits include: 
  • Peristaltic pump bulk transfer at its best
  • Highest flow rate
  • Flow ranges from 0.3 to 42 LPM
Our B/T Series pumps simply make your job more efficient. And with their heavy-duty motors and excellent hazard protection, they’re built to last. 
Masterflex B/T Series Pump Components
Reimage fluid handling
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
Masterflex B/T® MasterSense™ BioPharma Pump, EtherNet/IP™ and Profibus Network-Compatible, 11 to 321 rpm; 90 to 260 VAC
A broad flow range, from 0.65 to 42 LPM (0.17 to 11.1 GPM), for integration into your higher-flow bioprocess applications. Pump head accepts B/T PerfectPosition® pump tubing sizes B/T 87 and B/T 91 for repeatable and reliable performance. Open-head sensor technology stops the drive motor when pump head is opened.
Learn more
Custom services and tools
Masterflex Peristaltic Pump Series — Shop by flow range
Need more information or help selecting the perfect product for your application?
Our experts are ready to help.
Contact us