Masterflex® Ismatec® Microflow Series Peristaltic Pumps
Microflow, high accuracy
Precise control at your fingertips, in your lab or process.
Microflow, high accuracy pumping, flow range 0.0001 to 365 mL/min
With flow rates from 0.0001 to 365 mL/min, Masterflex Ismatec microflow pumps with low flow give you that precise control.
Ismatec microflow pumps are ideal for: 
  • Autoanalysis 
  • Flow cell 
  • Sipper 
  • OEM 
  • Low flow feed and transfer applications  
They offer the latest technology, including MasterflexLive, easy control and tubing changes, and peristaltic pumps' low maintenance and cost-effectiveness. Ismatec pumps accept Ismatec pump tubing for clean fluid paths. Tubing changes are quick and easy, with several available tubing formulations to ensure compatibility with your fluid.
Masterflex Ismatec Microflow Series Pump & Pump Tubing
Masterflex Ismatec Pumps Features and Benefits
Masterflex Ismatec Pumps benefits include:
  • Flow rates as low as 0.0001 mL/min
  • High-accuracy numerical, digital, or programmable speed control
  • Tubing is the only wetted part— Packed with advanced technology, most drives are MasterflexLive® enabled
  • Versatile- single, dual, multichannel head options
  • Most drives have a 2-year warranty— ISO 9000 Registered Supplier
  • Other Pump Technologies
Masterflex Mastersense BT BioPharma Touchscreen
Continous Mode
Masterflex Mastersense BT BioPharma Touchscreen
Time Mode
Masterflex Mastersense BT BioPharma Touchscreen
Volume Mode
Masterflex Mastersense BT BioPharma Touchscreen
Remote Mode
Masterflex Mastersense BT BioPharma Touchscreen
Tube Calibration
Our intuitive touchscreen controls feature
  • Batch control, interval/off time in volume and time mode
  • Flexible operational control via screen and analog remote control (monitoring via MasterflexLive)
  • Network connection via standard Ethernet (RJ45) or connectivity via Wi-Fi
  • USB connection allows for downloadable firmware updates to be loaded into the pump
  • Advanced connectivity for remote control includes EtherNet/IP and Serial via USB
Featured Easy-to-Use Ismatec Pumps
In addition to other ISMATEC® Peristaltic Pumps, our featured products include:
Masterflex® Ismatec® Reglo Digital Miniflex Pumps, Avantor®
Versatile pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and retrieve commonly used dispense programs for rapid setup
  • Anti-drip and speed ramping available in Volume dispense mode – ensure accuracy and minimize sample loss
  • Miniflex pump head accepts continuous tubing for clean, uninterrupted fluid paths
  • Three-level user access – gives you the option of controlling who can operate the pump and at what level
MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps; get real-time notifications of pump parameters – including speed, flow rate, dispense volume, and more. You can monitor critical processes running 24/7 without needing onsite personnel while push notifications alert you to operating conditions and error messages.
Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of the drive; USB connection allows for downloadable firmware updates to be loaded into the pump.
The Miniflex pump head features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long-wearing for smooth operation and long life.
Masterflex® Ismatec® IPC High-Accuracy Multichannel Peristaltic Pumps, Avantor®
The next generation Masterflex Ismatec IPC is a flexible and versatile multichannel pump system that offers accuracy and repeatability in up to 24 synchronous channels for greater throughput and performance. Perfect for auto-analysis, reactor studies, cell line seeding, and research.
  • Planetary drive with 8 stainless steel rollers reduces pulsation, increases flow rate accuracy, and lengthens tubing life
  • New intuitive 12.7 cm (5") capacitive TFT/RGB touchscreen UI for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or remote control (analog remote control, serial, or EtherNet/IP)
  • Three-level user access – gives you the option of controlling who can operate the pump and at what level
  • Pump lets you save and retrieve up to 50 commonly used dispense programs for rapid set up
  • Choose continuous run, volume and time dispense modes; including batch control and interval/off time
These Ismatec® IPC multichannel pumps feature planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. The intuitive touchscreen display shows flow rate or RPM, allows for easy calibration, and features configurable volume or time dispense modes. Volumetric dispensing features a unique roller positioning system to ensure repeatability.
Mix and match tubing sizes and formulations across the channels to suit your application. Included Click’n’go cartridges feature automatic occlusion for simplified use, fewer tubing changes, and reproducible results.
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Independently control flow and direction of each channel via keypad or your PC.
  • Flow rate 0.001 to 43 ml/min per channel - flow rate depends on drive rpm, rollers, and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer—expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensing with the capability of bidirectional flow in each channel. Plus, independent channel calibration minimizes tube-to-tube differences, resulting in high calibration accuracy.
Masterflex® Ismatec® Reglo Digital Multichannel Pumps, Avantor®
Versatile multichannel pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Anti-drip and speed ramping available in Volume dispense mode – ensure accuracy and minimize sample loss
  • Cassette pump models accept 3-stop tubing for easy loading and secure retention
  • Three-level user access – gives you the option of controlling who can operate the pump and at what level
MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps; get real-time notifications of pump parameters – including speed, flow rate, dispense volume, and more. You can monitor critical processes running 24/7 without the need for onsite personnel while push notifications alert you to operating conditions and error messages.
Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive; USB connection allows for downloadable firmware updates to be loaded into the pump.
Tubing cassettes (included) accept 3-stop tubing and are easily snapped in/out for quick tubing changes. Accepted tubing ranges from 0.13 to 3.17 mm ID for a wide flow range.
Masterflex® Ismatec® Microflex Variable-Speed Pump Systems, Avantor®
Easy-to-use low-flow pumps feature reversible, remote start/stop, and rapid tubing changes.
  • Polyphenylene sulfide (PPS), acetal, and copolyester pump head with four stainless steel (SS) rollers
  • Pumps accept continuous lengths of tubing for a clean, uninterrupted flow path with no fittings.
  • Separate single-turn speed control and CW/off/CCW switch with green LED power indicator—maintain speed when turning the pump on/off or in reverse direction.
  • Reversible permanent magnet DC motor; purge fluid before or after pumping; pump fluid in either direction.
  • Remote control connections on the back of pump; start/stop pump with a contact closure.
These easy-to-use pumps feature reversible pumping, remote control capabilities, and rapid tubing changes. Ideal for consistent fluid delivery in analyzer and other low-flow laboratory pumping applications. The quick-loading Microflex pump head has average fixed occlusion for tubing; no occlusion adjustments required.
Custom services and tools
Need more information or help selecting the perfect product for your application?
Our experts are ready to help.
Contact us