Oxford Nanopore Technologies
The affordable, pocket-sized MinION and MinION Mk1C devices, which can deliver tens of gigabases of data in a single run, make sequencing accessible to any researcher across a wide range of applications.
Benefits of nanopore sequencing technology include:
  • Any length read — from short to ultra-long (>4 Mb)
  • Streamlined workflows — library prep in as little as 10 minutes
  • Sequence native DNA/RNA — eliminate amplification-bias and directly identify epigenetic modifications (e.g. methylation)
  • Real-time data streaming — immediate access to actionable results
  • Portable, affordable devices — sequence anywhere