Rotary Evaporators
Rotary Evaporators
Rotary evaporators are used for the efficient and gentle removal of solvents from samples by evaporation, under reduced atmospheric pressure, thus allowing the solvents to evaporate faster at low temperatures.
Check out the range!
Rotary evaporators, Heidolph
The Hei-VAP Core deliberately forgoes complex menus and automatic programming. Instead it focuses on simple operation, direct access to parameters and visual activity displays.
EXPLORE MORE