Automated Nucleic Acid PurificationFeatured Solutions![]() Omega Bio-tek Mag-Bind kits for automated nucleic acid purification solutions. Compatible with most open-programming robotic platforms. Additional Resources![]() AcroPrep™ Advance 96-well long tip filter plate for Nucleic Acid Binding incorporates a silica-based quartz glass fiber media to allow for efficient binding of DNA and RNA, while providing smooth flow and rapid processing of samples. AcroPrep™ Advance 96-Well Filter Plates for Ultrafiltration incorporates Omega™ membrane and provide rapid, efficient separation of biomolecules. AcroPrep™ Advance 96-well long tip filter plates AcroPrep™ Advance ultrafiltration plates |