Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Masterflex® Ismatec® Pump Tubing, Viton®, 6.4 mm ID x 11.2 mm OD; 25 ft
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX06519-34EA 78700 INR InStock
MFLX06519-34
Masterflex® Ismatec® Pump Tubing, Viton®, 6.4 mm ID x 11.2 mm OD; 25 ft
Tubing Peristaltic Pump Tubing
VWR Item#MFLX06519-34
Your Organization's Price:
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Dimensions: 6.4 mm ID x 11.2 mm OD; 25 ft
This opaque black tubing is ideal for general lab use including acid and solvent transfer or high temperature applications.
Other Products In this Family
SPECIFICATIONS
Key Features
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Dimensions: 6.4 mm ID x 11.2 mm OD; 25 ft
More About This Item
This opaque black tubing is ideal for general lab use including acid and solvent transfer or high temperature applications.
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms