Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® EZ-Vision® In-gel Solution
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR® EZ-Vision®
N391-0.5MLEA 9972 INR InStock
N391-0.5ML N391-5MLDRP
VWR® EZ-Vision® In-gel Solution
Stains Electrophoresis Stains
EZ-Vision in-gel solution is a non mutagenic and non toxic fluorescent DNA dye used as an in-gel or post-run stain to visualise DNA in agarose gels.
  • Immediate results, easy and convenient
  • Excellent replacement for the often used, but hazardous, ethidium bromide
  • Simply added to molten agarose prior to gel casting, allowing for instant visualisation of DNA bands upon UV exposure, as an in-gel preparation for electrophoresis
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms