Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Mycoplasma treatment kit, Myco-1 & 2 set
Supplier: PanReac AppliChem
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
A8360.0010EA 0 INR InStock
APLIA8360.0010
Mycoplasma treatment kit, Myco-1 & 2 set
For the treatment of Mycoplasma-infected cells.
Myco-1 & 2 set contains Myco-1 (A5222) and Myco-2 (A5233). Myco-1 is based on the antibiotic Tiamulin, which is produced by the fungus Pleurotus mutilus. Myco-2 is based on Minocycline, a Tetracycline derivative. Myco-1 and Myco-2 are generally used sequentially in combination. Embryonic stem cells (ES cells) were successfully treated with the two reagents. No cytotoxic effects were observed.
Myco-1 & 2 set contains Myco-1 (A5222) and Myco-2 (A5233). Myco-1 is based on the antibiotic Tiamulin, which is produced by the fungus Pleurotus mutilus. Myco-2 is based on Minocycline, a Tetracycline derivative. Myco-1 and Myco-2 are generally used sequentially in combination. Embryonic stem cells (ES cells) were successfully treated with the two reagents. No cytotoxic effects were observed.
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms