Synonyms:
Peroxidisulphuric acid diammonium salt
, Ammonium persulphate
K833-100TABSEA
7719
INR
InStock
K833-100TABS
Ammonium peroxodisulphate (APS) 150 mg, tablets, Biotechnology Grade
Ammonium peroxodisulphate (APS)
Ammonium persulphate forms free radicals when dissolved in water, which initiate polymerisation of acrylamide solutions.
- Convenient tablet, easily prepared without weighing
- Fast and easy polymerisation of acrylamide mixtures
- Odour-free and non flammable
- Non hazardous shipping
Due to the instability of APS in water, a working solution must be prepared daily for optimum performance. A 10% APS solution can be easily be prepared, without weighing, by adding one 150 mg APS tablet to 1,5 ml of water.