VWR® BOD Bottles, with Glass Stoppers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
215-3154EA 0 INR
215-3154 215-3155
VWR® BOD Bottles, with Glass Stoppers
Bottles BOD Bottles
Borosilicate glass, 3,3.
BOD bottle with glass stopper manufactured from ASTM E438 Type 1 Class A.