Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Burettes, Glass Key Stopcock
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
612-4217EA 0 INR InStock
612-4217 612-4219 612-4226
VWR® Burettes, Glass Key Stopcock
Burettes
Borosilicate glass 3,3, class A.
With straight bore glass key stopcock, individual work certificate, serially numbered.
Accuracy as per class AS DIN 12700, ISO 385.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms