Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Burettes, PTFE Stopcock
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
612-4265EA 0 INR InStock
612-4265 612-4266 612-4267
VWR® Burettes, PTFE Stopcock
Burettes
Borosilicate glass 3,3, class AS.
With detachable PTFE stopcock, Schellbach stripe.
AS per DIN ISO 385 and USP standards.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms