Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

3,5-Dinitrobenzoic acid 20 g/l in ethanol Reag. Ph. Eur. 1031301

Supplier: VWR Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

87819.290EA 52651 INR InStock
87819.290
3,5-Dinitrobenzoic acid 20 g/l in ethanol Reag. Ph. Eur. 1031301
3,5-Dinitrobenzoic acid
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.


Formula: C₇H₄N₂O₆
MW: 212,12 g/mol
Melting Pt: 204 to 208 °C
Density: 1,683 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00007253
CAS Number: 99-34-3
EINECS: 202-751-1
UN: 1993
ADR: 3,II
Merck Index: 13,03304
Order Now

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms