Penicillin : Streptomycin 100X for tissue culture
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
K952-100MLEA
6563
INR
K952-100ML
Penicillin : Streptomycin 100X for tissue culture
Penicillin/Streptomycin
Pennicillin-Streptomycin 100X sterile solution for tissue culture applications. Inhibits cell wall synthesis and protein sythesis in gram-positive and gram-negative bacteria.
- 10 000 U/ml Penicillin, 10 mg/ml Streptomycin
- Endotoxin-tested, liquid, stable, antibiotic solution
- Targets gram-positive and gram-negative bacteria
CAS Number:
1406-05-9 (Penicillin) - 57-92-1 (Streptomycin) |
Specification Test Results
Antimicrobial Activity (1:100) | PASS |
Endotoxins (1:100) | 0.25 EU/mL |
Osmolality | REPORT |
pH @25 °C | REPORT |
USP Sterility | PASS |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...