Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
PerfeCTa® DNase I
Supplier: Quantabio
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
QUNT95150-100EA 1500 INR InStock
QUNT95150-100 QUNT95150-01K
PerfeCTa® DNase I
PerfeCTa® DNase I is a high purity, recombinant bovine DNase I preparation that is free of any contaminating RNases or proteases. It provides a simple and rapid solution to eliminate residual genomic DNA from total RNA preparations for expression profiling by reverse transcription quantitative PCR amplification (RT-qPCR) as well as other molecular biology applications.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms