These Cryo-Babies® and Cryo-Tags® polyolefin cryo-labels are specially top-coated to readily accept laser printing.
- Temperature resistant from −196 to +150 °C
- Chemically inert, resist most organic solvents and caustic agents
- Adhere to most plastics, glass and metals without cracking, peeling or degrading
Cryo-Babies® and Cryo-Tags® withstand freezer temperatures down to −196 °C (liquid and vapor phase nitrogen), boiling water baths (100 °C), autoclaving and moderate oven temperatures (150 °C).