Thermal transfer Cryo-Tags® labels
Supplier: Diversified Biotech
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Cryo-Tags®
DIVECRTH-2000EA 0 INR
DIVECRTH-2000
Thermal transfer Cryo-Tags® labels
Labels
These Cryo-Tags® polyester labels work with most thermal transfer printers to print popular barcode formats for fast and easy sample identification.
  • Temperature resistant from −196 to +80 °C
  • Freezable in liquid and vapour phase nitrogen
  • Solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
  • Resists degradation from scuffing, chemicals, moisture and temperature fluctuations
Print crisp, clear, professional quality labels.