Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Microinjectors, Recording Nanoject II™
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
DRUM3-000-205-RNEA 0 INR InStock
DRUM3-000-205-RN
Microinjectors, Recording Nanoject II™
The Recording Nanoject II™ microinjector was specifically developed to improve micro-volume injection techniques used to study connectivity and function of specific brain nuclei. Designed to be operated in conjunction with an extracellular amplifier, the integrated system can be used to reliably place small volumes into targeted areas. It is engineered to easily mount on popular stereotaxic instruments.
  • Enables high precision
  • Facilitates connection to an extracellular amplifier to allow recording of neural activity
  • Supplements stereotaxic coordinates in guiding the placement of small injections
  • Provides direct displacement of neuronal tracer or excitotoxin from the pipette
  • Can be used on stereotaxic equipment utilising 7,9 mm clamps
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms