Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Centrifuge tubes, conical bottom
Supplier: Brand
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
525-1253EA 0 INR InStock
BRND778313 BRND778713
Centrifuge tubes, conical bottom
DURAN® borosilicate glass 3.3.
  • Withstands forces up to 3500×g
  • Good temperature resistance
  • Good chemical resistance
Conical bottom, beaded rim, graduated. Volume and inscriptions are in high contrast white enamel.
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms