Syringes, interchangeable, Micro-Mate®
Supplier: Cadence Science
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Micro-Mate®
POPR5025 POPR5037 POPR5016 POPR5038 POPR5027
Syringes, interchangeable, Micro-Mate®
Syringes Syringes, Medical
These interchangeable syringes are manufactured to ensure the perfect mating of barrels and plungers. This Micro-Mate® interchangeable syringe of a particular size will fit any Micro-Mate® interchangeable barrel of that same size.
  • Made from borosilicate glass providing protection against thermal shock
  • Corrosion resistant
  • Precision shrinking ensures improved fit
  • Coloured plunger ends are fused around bottom beveled rim of plunger to facilitate dosage measurements
  • Graduations permanently fused into glass barrel
  • Reinforced flanges prevent syringe rolling