Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Capillary viscometers, Cannon-Fenske Opaque
Supplier: Fungilab
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
FUNGCV004-101EA 0 INR InStock
FUNGCV004-103 FUNGCV004-104 FUNGCV004-105 FUNGCV004-106 FUNGCV004-107 FUNGCV004-108 FUNGCV004-109 FUNGCV004-110 FUNGCV004-111 FUNGCV004-101 FUNGCV004-112 FUNGCV004-102
Capillary viscometers, Cannon-Fenske Opaque
Viscometers Capillary Viscometers
Cannon-Fenske Opaque viscometers (reverse flow type) for dark Newtonian liquids especially for liquids so dark in colour that cannot be seen in a Cannon-Fenske routine viscometer.
Used to study shearing stress and shearing rate.
Certifications: ASTM D-445-446, ASTM D2515, ISO 3104-3105 and UNE 400313
Delivery information: Supplied with the calibration certificate granted by a National Metrological Register with recognised international standards UKAS.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms