This combination ORP/Redox electrode with platinum wire sensor features a replaceable reference electrolyte gel cartridge and an integrated temperature sensor. The electrode has a 5-pin connector.
- Reference junction is resistant to clogging and easy to clean: Perfect for dirtier, more difficult samples where redox measurements are commonly made
- Replaceable gel reference electrolyte cartridge is a fast and easy maintenance solution that maximise the life of the electrode
- Rugged and durable: Designed to withstand the rigors of outdoor testing and frequent use
Compatible with Hach sensION systems.
Delivery information: Electrode is supplied with two replacement electrolyte gel cartridges and user manual.