Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Lactose MPN broth tubes
Supplier: Hach
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
HACH2101315EA 0 INR InStock
HACH2101315
Lactose MPN broth tubes
Liquid MPN media (broth) is used for conducting the Most Probable Number (MPN) method of estimating the number of viable microbiological organisms in a sample matrix.
  • Easy to use
  • Individually-packaged glass tubes
  • Manufactured and inspected in-house
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms