Phosphonate test kit, PN-10
Supplier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HACH2113302EA 0 INR
HACH2113302
Phosphonate test kit, PN-10
Environmental Test Kits
The phosphonate test kit comes with a 230 V, UV lamp which will digest and convert phosphonates into reactive phosphate.
  • Simple and fast colour disc method
  • Control phosphonate concentration
Delivery information: Kit includes colour disc, comparator box and tubes, volumetric flask, graduated cylinder, reaction bottle, reagents, UV lamp, 230 V power supply, UV safety goggles, instructions and carrying case.