Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Chemical Resistant Marker
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU95042-566EA 5721 INR InStock
VWRU95042-566
VWR®, Chemical Resistant Marker
Pens Permanent Markers
Precision ultrafine tip permanently marks slides and cassettes. Ink includes formalin and xylene. Note: Markers are not for use on smooth surfaces.
  • Ink is waterproof, smearproof, odorless and resistant to most chemicals
  • Used on frosted slides and tissue cassettes
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms