Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Dulbecco’s phosphate buffered saline (DPBS)
Supplier: Lonza
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
LONZ17-513F
Dulbecco’s phosphate buffered saline (DPBS)
DPBS is used as a buffer system to maintain cell culture media in the physiological pH range of 7,0 to 7,6.
It serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms