Combination pH electrodes, refillable, sensION™
Supplier: Hach Lange
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HACH5194000EA 0 INR
HACH5194000
Combination pH electrodes, refillable, sensION™
Standard combination pH electrodes with a refillable reference and built-in temperature sensor.
  • Easy to use and accurate
  • Rugged and durable: Designed to withstand the rigors of outdoor testing and frequent use
  • Flexible: Suitable for field and laboratory use
Compatible with Hach sensION meters.
Delivery information: Both electrodes are supplied with a small bottle of electrolyte refill solution and user manual.