Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Disposable Weighing Dishes, Aluminium, Crimped Sides with Tabs
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
25433-008 25433-085
25433-008EA 0 INR InStock
25433-008 25433-085
VWR®, Disposable Weighing Dishes, Aluminium, Crimped Sides with Tabs
Boats Weighing Boats
Aluminium.
  • Crimped sides for rigidity
  • No oil residue, no vinyl coating to create contamination problems for the user
  • Sturdy, with a flat bottom and slight taper for easy stacking
  • Dishes feature tabs for easy handling
These dishes are ideal for general-purpose weighing, dispensing, and storing applications. Dishes also function as evaporating dishes, parts containers, or dust covers.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms