Quartz-crystal stopwatch with dual display.
- Timing capacity: 9 hours, 59 minutes, 59 seconds and 99 hundredths
- Functions: Single action, time out, cumulative split, interval split and continuous roll-over timing
- Dual display shows cumulative and interval split
- Repeat timer provides repeat function of countdown alarm timing
- Eight recallable memories, plus fastest and slowest lap times
- Tough, water resistant ABS plastic casing
- Ideal for enzyme tests, viscosity measurements and all timed lab tests
Certifications: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: Supplied with lanyard, battery and certificate.