Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Luxmeter, digital, LU8500
Supplier: Industrial Physics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
SIMELU8500EA 0 INR InStock
SIMELU8500
Luxmeter, digital, LU8500
LU8500 luxmeter is a digital, handheld with large LCD display.
  • High accuracy and rapid response
  • Peak-hold function for tracing peak signals of light pulse with least duration 10 µs
  • Capable of selecting measuring mode in lux or fc scale alternatively
  • Measurement rate: 1,5 times per second
  • With maximum and minimum measurements
Delivery information: The instrument is delivered with a carrying case, instruction manual and battery.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms