Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Connectors, Tubing, Untapered
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
46600-088 46600-060 46600-062 46600-072 46600-148 46600-126 46600-058 46600-066
VWRU46600-088EA 0 INR InStock
VWRU46600-088 VWRU46600-060 VWRU46600-062 VWRU46600-072 VWRU46600-148 VWRU46600-126 VWRU46600-058 VWRU46600-066
VWR®, Connectors, Tubing, Untapered
Fittings and Connectors
PP, translucent.
  • Serrated arms to hold tubing tightly without slipping
  • Withstands flow up to 1,37 bar
  • High chemical and temperature resistance
  • Autoclavable
These connectors are suitable for making connections in a variety of tubing systems and vacuum lines. For general purpose use only.
Certifications: Non-ADI compliant.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms