Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Paddle Wheel Indicator
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU46600-114EA 0 INR InStock
VWRU46600-114
VWR®, Paddle Wheel Indicator
Flow Indicators
This free-spinning paddle wheel indicator is designed for liquid or gas flows.
  • Made in styrene-acrylonitrile (SAN)
  • Tapered arms connect to a variety of tubing sizes
  • Temperature and chemical resistant
Please note: Speed is proportional to flow rate.
Caution: Not for use with corrosive fluids or at temperatures above 85 °C.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms