VWR®, Automatic Burettes, with Stopcock
Supplier: VWR BRAND PRIVATE LABEL USA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17580-936 17579-959 17580-007
VWRU17580-936EA 0 INR
VWRU17580-936 VWRU17579-959 VWRU17580-007
VWR®, Automatic Burettes, with Stopcock
Burettes
Glass, class A, self-zeroing, graduated.
Amber glass burettes are mainly used for the light sensitive liquids.
Delivery information: Supplied with glass stopcock plug, bottle and rubber bulb.