Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Beakers, heavy-duty, low form, double scale
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWRU89000-222EA 0 INR InStock
VWRU89000-222
Beakers, heavy-duty, low form, double scale
Borosilicate glass, heavy-duty, with spout.
  • Durable matte finish marking area and a white graduated scale
  • Spout is designed for optimum performance
These beakers are with thick uniform walls and extra wall thickness built into the evenly tooled top rim. Heavy construction is intended to prolong life expectancy with harder than normal usage. Not recommended for use on hot plates where significant thermal gradients exist due to lack of agitation of contents or absence of a temperature control.
Certifications: Designed from ASTM Specification E960, Type II requirements
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms